Search Thermo Fisher Scientific
AbboMax
FIGURE: 1 / 1
Positive Control: Kidney Tissue. Cellular Location: Cell Membrane.
The human immunodeficiency virus type 1 (HIV-1) envelope glycoprotein is synthesized as a precursor glycoprotein, gp160, and is then processed into gp120 and gp41. HIV-1 infection is initiated by the binding of gp120 envelope glyco-protein to its cell receptor (CD4) and a coreceptor (CXCR4 or CCR5), followed by a series of conformational changes in the gp41 transmembrane subunit.