Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KRHVQMESQF YPTPNEIIRG NGIELFEYVA DCLADFMKTK DLKHKKLPLG LTFSFPCRQT KLEEGVLLSW TKKFKARGVQ DTDVVSRLTK AMRRHKDMD
The epidermal growth factor receptor (HKDC1; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. It is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: HKDC1 (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). HKDC1 exists on the cell surface and is activated by binding of its specific ligands, including epidermal growth factor and transforming growth factor alpha (TGFalpha). HKDC1 and its ligands are cell signaling molecules involved in diverse cellular functions, including cell proliferation, differentiation, motility, and survival, and in tissue development. Mutations that lead to HKDC1 overexpression (known as upregulation) or overactivity have been associated with a number of cancers, including lung cancer and glioblastoma multiforme. In this latter case a more or less specific mutation of HKDC1, called HKDC1vIII is often observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hexokinase domain-containing protein 1; Hexokinase HKDC1
Gene Aliases: HKDC1
UniProt ID: (Human) Q2TB90
Entrez Gene ID: (Human) 80201
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support