Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • HLA-A2 Antibodies

            Invitrogen

            HLA-A2 Monoclonal Antibody (BB7.2), APC, eBioscience™

            Advanced Verification
            4 Published Figures
            8 References
            View all (28) HLA-A2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 17-9876-42
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            341.70
            Special offer
            Online exclusive
            Ends: 01-Jul-2025
            402.00
            Save 60.30 (15%)
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite HLA-A2 Monoclonal Antibody (BB7.2), APC, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (4)
            • Advanced Verification (1)
            Application
            HLA-A2 Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 6

            HLA-A2 Antibody (17-9876-42) in Flow

            Staining of normal human peripheral blood cells with Mouse IgG2b K Isotype Control APC (Product # 17-4732-81) (blue histogram) or Anti-Human HLA-A2 APC (purple histogram). Cells in the lymphocyte gate were used for analysis.
            View Product

            HLA-A2 Antibody in Flow Cytometry (Flow)
            HLA-A2 Antibody in Flow Cytometry (Flow)
            HLA-A2 Antibody in Flow Cytometry (Flow)
            HLA-A2 Antibody in Flow Cytometry (Flow)
            HLA-A2 Antibody in Flow Cytometry (Flow)
            HLA-A2 Antibody

            Product Details

            17-9876-42

            Applications
            Tested Dilution
            Publications

            Flow Cytometry (Flow)

            5 µL (0.5 µg)/test
            View 8 publications 8 publications
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human

            Host/Isotype

            Mouse / IgG2b, kappa

            Recommended Isotype Control

            Mouse IgG2b kappa Isotype Control (eBMG2b), APC, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            BB7.2

            Conjugate

            APC
            • APC-eFluor 780
            • NovaFluor Blue 510
            • NovaFluor Blue 585
            • NovaFluor Blue 610-70S
            • NovaFluor Blue 660-120S
            • NovaFluor Blue 690
            • NovaFluor Red 710
            • NovaFluor Red 725
            • NovaFluor Yellow 570
            • NovaFluor Yellow 660
            • NovaFluor Yellow 730
            • NovaFluor Yellow 755
            • PE 
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-eFluor 710
            • Super Bright 436
            • Super Bright 600
            • Request custom conjugation

            Excitation/Emission Max

            651/660 nm View spectra spectra

            Form

            Liquid

            Concentration

            5 µL/Test

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2, with BSA

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_11149299

            Product Specific Information

            Description: This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with beta 2-microglobulin and is expressed on all nucleated cells.

            The BB7.2 monoclonal antibody has been reported to exhibit blocking activity, as well as recognize HLA-A28.

            Applications Reported: This BB7.2 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This BB7.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

            Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described.

            HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 8:21
            Loaded: 1.98%
            0:00
            Stream Type LIVE
            Remaining Time -8:21
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected

            This is a modal window.

            Beginning of dialog window. Escape will cancel and close the window.

            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family

            End of dialog window.

            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

            How to use the Panel Builder

            Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

            Related Products for

            Flow Cytometry
            • Additional Formats
            • Isotype Controls
            • Buffers FC
            • Viability Dyes FC
            • Compensation Beads FC
            • Labeling Kits FC
            Catalog #
            61-9876-42
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), PE-eFluor™ 610, eBioscience™
            Price (USD)
            442.00
            Unit Size :
            100 Tests
            Catalog #
            H070T03R04-A
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Red 710, eBioscience™
            Price (USD)
            430.00
            Unit Size :
            100 Tests
            Catalog #
            H070T03B04-A
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Blue 585, eBioscience™
            Price (USD)
            440.00
            Unit Size :
            100 Tests
            Catalog #
            H070T03Y01-A
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Yellow 570, eBioscience™
            Unit Size :
            100 Tests
            Catalog #
            H070T03B06-A
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Blue 610-70S, eBioscience™
            Unit Size :
            100 Tests
            Catalog #
            H070T03B01-A
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Blue 510, eBioscience™
            Unit Size :
            100 Tests
            Catalog #
            H070T03Y08-A
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Yellow 755, eBioscience™
            Unit Size :
            100 Tests
            Catalog #
            12-9876-42
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), PE, eBioscience™
            Unit Size :
            100 Tests
            Catalog #
            63-9876-42
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), Super Bright™ 600, eBioscience™
            Unit Size :
            100 Tests
            Catalog #
            H070T03Y07-A
            Invitrogen
            HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Yellow 730, eBioscience™
            Unit Size :
            100 Tests

            Bioinformatics

            Protein Aliases: A29 protein; antigen presenting molecule; DAQB-90C11.16; FLJ26655; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-Aw33.1; Human leukocyte antigen A; leucocyte antigen A; leukocyte antigen class I-A; MCH class I antigen; MHC 1; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; MHC1; truncated MHC class I antigen

            View more View less

            Gene Aliases: Aw-33; Aw-74; HLA-A; HLA-A11; HLA-A33; HLA-DQB1; HLA-DRB1; HLAA

            View more View less

            UniProt ID: (Human) Q95380

            View more View less

            Entrez Gene ID: (Human) 3105

            View more View less

            Function(s)

            protein binding beta-2-microglobulin binding peptide antigen binding poly(A) RNA binding major histocompatibility complex protein

            Process(es)

            antigen processing and presentation of peptide antigen via MHC class I antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent viral process antigen processing and presentation regulation of immune response interferon-gamma-mediated signaling pathway type I interferon signaling pathway
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-pkshg:80/100.66.78.18:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0