Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody recognizes an extracellular antigen of HLA-B7 antigen. Although highly specific, it can cross-react with HLA-B42 antigen.
HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDABP0067; class I MHC, HLA-B antigen; DAQB-90C11.16; epididymis secretory protein Li 83; FLJ26655; histocompatibility antigen; HLA -B; HLA B-40011; HLA B44; hla b7; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLA-B; HLA-B 1502; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B35; HLA-B35 antigen; HLA-Bw50 antigen; Human leukocyte antigen B; hypothetical protein; lecocyte antigen B; Leukocyte Antigen; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; MHC 1; MHC antigen; MHC class 1 antigen; MHC class I alpha chain; MHC class I anitgen; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen GN00104; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen SHCHA; MHC class I antigene; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27-HS; MHC HLA-B27d; MHC HLA-B39N; MHC1; truncated class I antigen; truncated MHC class I antigen
Gene Aliases: AS; B-4901; B-5001; Bw-47; Bw-50; HEL-S-83; HLA-B; HLA-B*45ZJ; HLA-B-3506; HLA-B-3905; HLA-B-5502; HLA-B-5602; HLA-B15; HLA-B39; HLA-B49; HLA-B50; HLA-B55; HLA-B59; HLA-B61; HLA-Cw; HLA-DRB1; HLAB; SPDA1
UniProt ID: (Human) Q9GJF0
Entrez Gene ID: (Human) 3106
Molecular Function:
major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support