Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene belongs to the HLA class II alpha chain family. The encoded protein forms a heterodimer with a class II beta chain. It is located in intracellular vesicles and plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells and are used to present antigenic peptides on the cell surface to be recognized by CD4 T-cells.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DC-1 alpha chain; DC-alpha; DQ alpha 2; DX alpha chain; HLA class II histocompatibility antigen, DQ alpha 1 chain; HLA class II histocompatibility antigen, DQ alpha 2 chain; HLA class II histocompatibility antigen, DQ(6) alpha chain; HLA class II histocompatibility antigen, DQ(W3) alpha chain; HLA-DCA; HLA-DQA1; leucocyte antigen DQA1; leukocyte antigen alpha chain; MHC class II antigen; MHC class II DQA1; MHC class II DQA2; MHC class II HLA-D alpha glycoprotein; MHC class II HLA-DQ-alpha-1; MHC class II surface glycoprotein; MHC HLA-DQ alpha
Gene Aliases: CD; CELIAC1; DQ-A1; DX-ALPHA; GSE; HLA-DQA; HLA-DQA1; HLA-DQA2; HLA-DXA
UniProt ID: (Human) P01909, (Human) P01906
Entrez Gene ID: (Human) 3117, (Human) 3118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support