Product References
TNF-α activates RELA expression via TNFRSF1B to upregulate OPA1 expression and inhibit chondrogenic differentiation of human adipose stem cells.
Journal of orthopaedic surgery and research
Guo J,Ye W,Wu X,Huang H,Li B,Sun Z,Ren Z,Yang Z
12-9956-42 was used in Flow cytometry/Cell sorting to show that the presence of TNF-α led to increased expression of TNFα receptor 2 (TNFRSF1B) and RELA during chondrogenic differentiation of hADSCs.
Tue Jun 13 00:00:00 UTC 2023
WT1 Pulsed Human CD141+ Dendritic Cell Vaccine Has High Potential in Solid Tumor-Targeted Immunotherapy.
International journal of molecular sciences
Cho SY,Jeong SM,Jeon YJ,Yang SJ,Hwang JE,Yoo BM,Kim HS
12-9956-42 was used in Flow cytometry/Cell sorting to develop a DC vaccine named CellgramDC-WT1 (CDW).
Thu Jan 12 00:00:00 UTC 2023
Coordinated glucocorticoid receptor and MAFB action induces tolerogenesis and epigenome remodeling in dendritic cells.
Nucleic acids research
Morante-Palacios O,Ciudad L,Micheroli R,de la Calle-Fabregat C,Li T,Barbisan G,Houtman M,Edalat SG,Frank-Bertoncelj M,Ospelt C,Ballestar E
12-9956-42 was used in Flow cytometry/Cell sorting to investigate epigenetic and transcriptomic changes implicated with dendritic cell differentiation with glucocorticoids.
Tue Jan 11 00:00:00 UTC 2022
Endothelial dysfunction markers and immune response indices in cosmonauts' blood after long-duration space flights.
NPJ microgravity
Kuzichkin DS,Nichiporuk IA,Zhuravleva OA,Markin AA,Rykova MP,Zhuravleva TV,Sadova AA,Kutko OV,Shmarov VA,Ponomarev SA
12-9956-42 was used in Flow cytometry/Cell sorting to investigat the markers of endothelial activation and damage (plasma concentrations of soluble thrombomodulin fraction (sTM), von Willebrand factor (vWF), highly sensitive C-reactive protein (hs-CRP)), as well as the level of D-dimer and compared them to the immunological parameters characterizing the state of human humoral and cellular immunity.
Wed Nov 02 00:00:00 UTC 2022
Activated CD4+ T cells and CD14hiCD16+ monocytes correlate with antibody response following influenza virus infection in humans.
Cell reports. Medicine
Wong SS,Oshansky CM,Guo XJ,Ralston J,Wood T,Reynolds GE,Seeds R,Jelley L,Waite B,Jeevan T,Zanin M,Widdowson MA,Huang QS,Thomas PG,Webby RJ
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Tue Apr 20 00:00:00 UTC 2021
The role of irreversible electroporation in promoting M1 macrophage polarization via regulating the HMGB1-RAGE-MAPK axis in pancreatic cancer.
Oncoimmunology
He C,Sun S,Zhang Y,Xie F,Li S
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Thu Mar 11 00:00:00 UTC 2021
Transplantation of human amniotic epithelial cells promotes morphological and functional regeneration in a rat uterine scar model.
Stem cell research & therapy
Fan Y,Sun J,Zhang Q,Lai D
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Wed Mar 24 00:00:00 UTC 2021
Lung transplantation for patients with severe COVID-19.
Science translational medicine
Bharat A,Querrey M,Markov NS,Kim S,Kurihara C,Garza-Castillon R,Manerikar A,Shilatifard A,Tomic R,Politanska Y,Abdala-Valencia H,Yeldandi AV,Lomasney JW,Misharin AV,Budinger GRS
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Wed Dec 16 00:00:00 UTC 2020
Gene expression network analyses during infection with virulent and avirulent Trypanosoma cruzi strains unveil a role for fibroblasts in neutrophil recruitment and activation.
PLoS pathogens
Oliveira AER,Pereira MCA,Belew AT,Ferreira LRP,Pereira LMN,Neves EGA,Nunes MDCP,Burleigh BA,Dutra WO,El-Sayed NM,Gazzinelli RT,Teixeira SMR
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Sat Aug 01 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
Characterization of HLA-G Regulation and HLA Expression in Breast Cancer and Malignant Melanoma Cell Lines upon IFN-γ Stimulation and Inhibition of DNA Methylation.
International journal of molecular sciences
Jørgensen N,Sayed A,Jeppesen HB,Persson G,Weisdorf I,Funck T,Hviid TVF
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Wed Jun 17 00:00:00 UTC 2020
Connecting METTL3 and intratumoural CD33+ MDSCs in predicting clinical outcome in cervical cancer.
Journal of translational medicine
Ni HH,Zhang L,Huang H,Dai SQ,Li J
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Thu Oct 15 00:00:00 UTC 2020
New GMP manufacturing processes to obtain thermostable HIV-1 gp41 virosomes under solid forms for various mucosal vaccination routes.
NPJ vaccines
Amacker M,Smardon C,Mason L,Sorrell J,Jeffery K,Adler M,Bhoelan F,Belova O,Spengler M,Punnamoottil B,Schwaller M,Bonduelle O,Combadière B,Stegmann T,Naylor A,Johnson R,Wong D,Fleury S
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Fri Dec 17 00:00:00 UTC 2021
Targeting the mTOR pathway uncouples the efficacy and toxicity of PD-1 blockade in renal transplantation.
Nature communications
Esfahani K,Al-Aubodah TA,Thebault P,Lapointe R,Hudson M,Johnson NA,Baran D,Bhulaiga N,Takano T,Cailhier JF,Piccirillo CA,Miller WH
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Thu Oct 17 00:00:00 UTC 2019
STAT3 Inhibition Combined with CpG Immunostimulation Activates Antitumor Immunity to Eradicate Genetically Distinct Castration-Resistant Prostate Cancers.
Clinical cancer research : an official journal of the American Association for Cancer Research
Moreira D,Adamus T,Zhao X,Su YL,Zhang Z,White SV,Swiderski P,Lu X,DePinho RA,Pal SK,Kortylewski M
12-9956 was used in Flow cytometry/Cell sorting to show a novel strategy combining STAT3 inhibition with Toll-like Receptor 9 (TLR9) stimulation to unleash immune response against prostate cancers regardless of the genetic background.
Sat Dec 01 00:00:00 UTC 2018
Comprehensive characterization of chorionic villi-derived mesenchymal stromal cells from human placenta.
Stem cell research & therapy
Ventura Ferreira MS,Bienert M,Müller K,Rath B,Goecke T,Opländer C,Braunschweig T,Mela P,Brümmendorf TH,Beier F,Neuss S
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2018
Large-Scale Human Dendritic Cell Differentiation Revealing Notch-Dependent Lineage Bifurcation and Heterogeneity.
Cell reports
Balan S,Arnold-Schrauf C,Abbas A,Couespel N,Savoret J,Imperatore F,Villani AC,Vu Manh TP,Bhardwaj N,Dalod M
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Tue Aug 14 00:00:00 UTC 2018
HDAC inhibition potentiates immunotherapy in triple negative breast cancer.
Oncotarget
Terranova-Barberio M,Thomas S,Ali N,Pawlowska N,Park J,Krings G,Rosenblum MD,Budillon A,Munster PN
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Tue Dec 26 00:00:00 UTC 2017
Gut-homing Δ42PD1+Vδ2 T cells promote innate mucosal damage via TLR4 during acute HIV type 1 infection.
Nature microbiology
Cheung AKL,Kwok HY,Huang Y,Chen M,Mo Y,Wu X,Lam KS,Kong HK,Lau TCK,Zhou J,Li J,Cheng L,Kiat Lee B,Peng Q,Lu X,An M,Wang H,Shang H,Zhou B,Wu H,Xu A,Yuen KY,Chen Z
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Sun Oct 01 00:00:00 UTC 2017
Surrogate in vitro activation of innate immunity synergizes with interleukin-7 to unleash rapid antigen-driven outgrowth of CD4+ and CD8+ human peripheral blood T-cells naturally recognizing MUC1, HER2/neu and other tumor-associated antigens.
Oncotarget
Pathangey LB,McCurry DB,Gendler SJ,Dominguez AL,Gorman JE,Pathangey G,Mihalik LA,Dang Y,Disis ML,Cohen PA
Tue Feb 14 00:00:00 UTC 2017
Allostimulatory capacity of conditionally immortalized proximal tubule cell lines for bioartificial kidney application.
Scientific reports
Mihajlovic M,van den Heuvel LP,Hoenderop JG,Jansen J,Wilmer MJ,Westheim AJF,Allebes WA,Stamatialis D,Hilbrands LB,Masereeuw R
12-9956 was used in Flow cytometry/Cell sorting to assess the immunosafety of conditionally immortalized proximal tubule cell lines for use in bioartificial kidneys, specifically by investigating their allogeneic efects.
Wed Aug 02 00:00:00 UTC 2017
FOXP1 suppresses immune response signatures and MHC class II expression in activated B-cell-like diffuse large B-cell lymphomas.
Leukemia
Brown PJ,Wong KK,Felce SL,Lyne L,Spearman H,Soilleux EJ,Pedersen LM,Møller MB,Green TM,Gascoyne DM,Banham AH
12-9956 was used in Flow cytometry/Cell sorting to identify FOXP1 as a novel regulator of genes targeted by the class II MHC transactivator CIITA.
Tue Mar 01 00:00:00 UTC 2016
Formin-generated actomyosin arcs propel T cell receptor microcluster movement at the immune synapse.
The Journal of cell biology
Murugesan S,Hong J,Yi J,Li D,Beach JR,Shao L,Meinhardt J,Madison G,Wu X,Betzig E,Hammer JA
12-9956 was used in Flow cytometry/Cell sorting to reveal the derivation and organisation of actin arcs in the immunological synapse.
Mon Nov 07 00:00:00 UTC 2016
Lack of T Cell Response to iPSC-Derived Retinal Pigment Epithelial Cells from HLA Homozygous Donors.
Stem cell reports
Sugita S,Iwasaki Y,Makabe K,Kimura T,Futagami T,Suegami S,Takahashi M
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Tue Oct 11 00:00:00 UTC 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
Published figure using HLA-DR monoclonal antibody (Product # 12-9956-42) in Flow Cytometry
Tue Aug 09 00:00:00 UTC 2016
Pentraxin-3 regulates the inflammatory activity of macrophages.
Biochemistry and biophysics reports
Shiraki A,Kotooka N,Komoda H,Hirase T,Oyama JI,Node K
12-9956 was used in Immunocytochemistry to investigate the function of pentraxin-3 in macrophages in vitro.
Tue Mar 01 00:00:00 UTC 2016
CX₃CR1⁺ mononuclear phagocytes support colitis-associated innate lymphoid cell production of IL-22.
The Journal of experimental medicine
Longman RS,Diehl GE,Victorio DA,Huh JR,Galan C,Miraldi ER,Swaminath A,Bonneau R,Scherl EJ,Littman DR
12-9956 was used in Flow cytometry/Cell sorting to study the role of CX3CR1+ mononuclear phagocytes in the development of colitis and disease progression following Citrobacter rodentium infection.
Mon Jul 28 00:00:00 UTC 2014
The CD14+CD16+ inflammatory monocyte subset displays increased mitochondrial activity and effector function during acute Plasmodium vivax malaria.
PLoS pathogens
Antonelli LR,Leoratti FM,Costa PA,Rocha BC,Diniz SQ,Tada MS,Pereira DB,Teixeira-Carvalho A,Golenbock DT,Gonçalves R,Gazzinelli RT
12-9956 was used in Flow cytometry/Cell sorting to study inflammatory monocyte subsets displaying increased mitochondrial activity and effector function during malaria.
Mon Sep 01 00:00:00 UTC 2014
Incomplete normalization of regulatory t-cell frequency in the gut mucosa of Colombian HIV-infected patients receiving long-term antiretroviral treatment.
PloS one
Rueda CM,Velilla PA,Chougnet CA,Rugeles MT
12-9956 was used in Flow cytometry/Cell sorting to assess the frequency and phenotype of regulatory T-cells within CMV and HIV patients on different treatment regimes.
Tue Sep 23 00:00:00 UTC 2014
Stem cell microvesicles transfer cystinosin to human cystinotic cells and reduce cystine accumulation in vitro.
PloS one
Iglesias DM,El-Kares R,Taranta A,Bellomo F,Emma F,Besouw M,Levtchenko E,Toelen J,van den Heuvel L,Chu L,Zhao J,Young YK,Eliopoulos N,Goodyer P
12-9956 was used in Flow cytometry/Cell sorting to show that human mesenchymal stem cells, from amniotic fluid or bone marrow, reduce pathologic cystine accumulation in co-cultured CTNS mutant fibroblasts or proximal tubular cells from cystinosis patients.
Tue Apr 23 00:00:00 UTC 2013
HIV-induced T-cell activation/exhaustion in rectal mucosa is controlled only partially by antiretroviral treatment.
PloS one
Rueda CM,Velilla PA,Chougnet CA,Montoya CJ,Rugeles MT
12-9956 was used in Flow cytometry/Cell sorting to investigate the effect of chronic HIV infection on the immune cell phenotype.
Mon Jun 04 00:00:00 UTC 2012
Dendritic cells derived from hemozoin-loaded monocytes display a partial maturation phenotype that promotes HIV-1 trans-infection of CD4+ T cells and virus replication.
Journal of immunology (Baltimore, Md. : 1950)
Diou J,Tardif MR,Barat C,Tremblay MJ
12-9956 was used in Flow cytometry/Cell sorting to investigate the effect of malarial pigment hemozoin on HIV-1 transmission, showing that hemozoid-loaded monocytes promote HIV-1 trans-infection and replication.
Mon Mar 15 00:00:00 UTC 2010
Down-regulation of MHC class II expression through inhibition of CIITA transcription by lytic transactivator Zta during Epstein-Barr virus reactivation.
Journal of immunology (Baltimore, Md. : 1950)
Li D,Qian L,Chen C,Shi M,Yu M,Hu M,Song L,Shen B,Guo N
12-9956 was used in Flow cytometry/Cell sorting to indicate that Zta may act as an inhibitor of the MHC class II pathway, suppressing CIITA transcription.
Sun Feb 15 00:00:00 UTC 2009
Immunological aspects of REIC/Dkk-3 in monocyte differentiation and tumor regression.
International journal of oncology
Watanabe M,Kashiwakura Y,Huang P,Ochiai K,Futami J,Li SA,Takaoka M,Nasu Y,Sakaguchi M,Huh NH,Kumon H
12-9956-42 was used in Flow cytometry/Cell sorting to study that recombinant REIC/Dkk-3 protein induced the differentiation of human CD14+ monocytes into a novel cell type.
Sun Mar 01 00:00:00 UTC 2009
A distinct Toll-like receptor repertoire in human tonsillar B cells, directly activated by PamCSK, R-837 and CpG-2006 stimulation.
Immunology
Månsson A,Adner M,Höckerfelt U,Cardell LO
12-9956 was used in Flow cytometry/Cell sorting to emphasize the unique ability of B cells to respond directly, via TLRs, to a certain range of microbial structures, further stressing the importance of TLRs in our defence against invading pathogens.
Tue Aug 01 00:00:00 UTC 2006
Expansion of toll-like receptor 9-expressing B cells in active systemic lupus erythematosus: implications for the induction and maintenance of the autoimmune process.
Arthritis and rheumatism
Papadimitraki ED,Choulaki C,Koutala E,Bertsias G,Tsatsanis C,Gergianaki I,Raptopoulou A,Kritikos HD,Mamalaki C,Sidiropoulos P,Boumpas DT
12-9956 was used in Flow cytometry/Cell sorting to examine the expression and function of various TLRs in subpopulations of peripheral blood mononuclear cells of patients with systemic lupus erythematosus.
Wed Nov 01 00:00:00 UTC 2006