Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Peptide sequence: PWVEQEGPQY WEWTTGYAKA NAQTDRVALR NLLRRYNQSE AGSHTLQGMN
Sequence homology: Human: 100%; Pig: 100%
HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDA12; HLA class I histocompatibility antigen, alpha chain F; HLA class I molecule; HLA F antigen; Leukocyte antigen F; MHC class I antigen F; MHC class Ib antigen
Gene Aliases: CDA12; HLA-5.4; HLA-CDA12; HLA-F; HLAF
UniProt ID: (Human) P30511
Entrez Gene ID: (Human) 3134
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support