Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGNGNAAA TAEENSPKMR VIRVGTRKSQ LARIQTDSVV ATLKASYPGL QFEIIAMSTT GDKILDTALS KIGEKSLFTK ELEHALEKNE VDLVVHSLKD LPTVLPPGFT IGAICKRENP HDAVVFHPKF VGKTLETLPE KSVVGTSSLR RAAQLQRKFP HLEFRSIRGN LNTRLRKLDE QQEFSAIILA TAGLQRMGWH NRVGQILHPE ECMYAVGQGA LGVEVRAKDQ DILDLVGVLH DPETLLRCIA ERAFLRHLEG GCSVPVAVHT AMKDGQLYLT GGVWSLDGSD SIQETMQATI HVPAQHEDGP EDDPQLVGIT ARNIPRGPQL AAQNLGISLA NLLLSKGAKN ILDVARQLND AH (1-361 aa encoded by BC000520)
PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternative name: porphobilinogen deaminase; HEM3; HMBS; Hydroxymethylbilane synthase; PBG D; PBG-D; Porphobilinogen deaminase; porphyria, acute; Chester type; Pre-uroporphyrinogen synthase; uroporphyrinogen I synthase; uroporphyrinogen I synthetase
Gene Aliases: hemC; HMBS; PBG-D; PBGD; PORC; T25658; UPS; URO-S; Uros1
UniProt ID: (Human) P08397, (Mouse) P22907
Entrez Gene ID: (Human) 3145, (Mouse) 15288, (Rat) 25709
Molecular Function: deaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support