Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MAYDDSVKKE DCFDGDHTFE DIGLAAGRSQ REKKRSYKDF LREEEEIAAQ VRNSSKKKLK DSELYFLGTD THKK
High mobility group (HMG) proteins are nonhistone chromosomal proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High mobility group protein 2-like 1; high-mobility group protein 2-like 1; HMG box domain containing 4; HMG box-containing protein 4; HMG domain-containing protein 4; Protein HMGBCG; Protein THC211630
Gene Aliases: HMG2L1; HMGBCG; HMGXB4; THC211630
UniProt ID: (Human) Q9UGU5
Entrez Gene ID: (Human) 10042
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support