Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAKGDPKKPK GKMSAYAFFV QTCREEHKKK NPEVPVNFAE FSKKCSERWK TMSGKEKSKF DEMAKADKVR YDREMKDYGP AKGGKKKKDP NAPKRPPSGF FLFCSEFRPK IKSTNPGISI GDVAKKLGEM WNNLNDSEKQ PYITKAAKLK EKYEKDVADY KSKGKFDGAK GPAKVARKKV EEEDEEEEEE EEEEEEEEDE
HMGB3 belongs to the high mobility group protein superfamily. Like HMG1 and HMG2, HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High mobility group protein 2a; High mobility group protein 4; High mobility group protein B3; high-mobility group (nonhistone chromosomal) protein 4; HMG-2a; HMG-4; MGC90319
Gene Aliases: HMG-2a; HMG-4; HMG2A; HMG4; HMGB3
UniProt ID: (Human) O15347
Entrez Gene ID: (Human) 3149
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support