Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MPGSLPLNAE ACWPKDVGIV ALEIYFPSQY VDQAELEKYD GVDAGKYTIG LGQAKMGFCT DREDINSLCM TVVQNLMERN NLSYDCIGRL EVGTETIIDK SKSVKTNLMQ LFEESGNTDI EGIDTTNACY GGTAAVFNAV NWIESSSWDG RYALVVAGDI AVYATGNARP TGGVGAVALL IGPNAPLIFE RGLRGTHMQH AYDFYKPDML SEYPIVDGKL SIQCYLSALD RCYSVYCKKI HAQWQKEGND KDFTLNDFGF MIFHSPYCKL VQKSLARMLL NDFLNDQNRD KNSIYSGLEA FGDVKLEDTY FDRDVEKAFM KASSELFSQK TKASLLVSNQ NGNMYTSSVY GSLASVLAQY SPQQLAGKRI GVFSYGSGLA ATLYSLKVTQ DATPGSALDK ITASLCDLKS RLDSRTGVAP DVFAENMKLR EDTHHLVNYI PQGSIDSLFE GTWYLVRVDE KHRRTYARRP TPNDDTLDEG VGLVHSNIAT EHIPSPAKKV PRLPATAAEP EAAVISNGEH
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); HMG-CoA synthase; Hydroxymethylglutaryl-CoA synthase, cytoplasmic
Gene Aliases: HMGCS; HMGCS1
UniProt ID: (Human) Q01581
Entrez Gene ID: (Human) 3157
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support