Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPKRKAEGD AKGDKAKVKD EPQRRSARLS AKPAPPKPEP KPKKAPAKKG EKVPKGKKGK ADAGKEGNNP AENGDAKTDQ AQKAEGAGDA K (1-90 aa encoded by BC014644)
The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High mobility group nucleosome-binding domain-containing protein 2; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17
Gene Aliases: HMG17; HMGN2
UniProt ID: (Human) P05204
Entrez Gene ID: (Human) 3151
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support