Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Binds to nucleosomes, regulating chromatin structure and consequently, chromatin-dependent processes such as transcription, DNA replication and DNA repair. Affects both insulin and glucagon levels and modulates the expression of pancreatic genes involved in insulin secretion. Regulates the expression of the glucose transporter SLC2A2 by binding specifically to its promoter region and recruiting PDX1 and additional transcription factors. Regulates the expression of SLC6A9, a glycine transporter which regulates the glycine concentration in synaptic junctions in the central nervous system, by binding to its transcription start site. May play a role in ocular development and astrocyte function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: High mobility group nucleosome-binding domain-containing protein 3; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP-7
Gene Aliases: HMGN3; PNAS-24; PNAS-25; TRIP7
UniProt ID: (Human) Q15651
Entrez Gene ID: (Human) 9324
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support