Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASSMRSLF SDHGKYVESF RRFLNHSTEH QCMQEFMDKK LPGIIGRIGD TKSEIKILSI GGGAGEIDLQ ILSKVQAQYP GVCINNEVVE PSAEQIAKYK ELVAKTSNLE NVKFAWHKET SSEYQSRMLE KKELQKWDFI HMIQMLYYVK DIPATLKFFH SLLGTNAKML IIVVSGSSGW DKLWKKYGSR FPQDDLCQYI TSDDLTQMLD NLGLKYECYD LLSTMDISDC FIDGDENGDL LWDFLTETCN FNATAPPDLR AELGKDLQEP EFSAKKEGKV LFNNTLSFIV IEA (1-292 aa encoded by BC020677)
In mammals, histamine is metabolized by two major pathways: N(tau)-methylation via histamine N-methyltransferase and oxidative deamination via diamine oxidase. HNMT is the first enzyme which is found in the cytosol and uses S-adenosyl-L-methionine as the methyl donor. In the mammalian brain, the neurotransmitter activity of histamine is controlled by N(tau)-methylation as diamine oxidase is not found in the central nervous system. A common genetic polymorphism affects the activity levels of this gene product in red blood cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histamine N-methyltransferase; HMT
Gene Aliases: 1500031F01Rik; AI788969; HMT; HNMT; HNMT-S1; HNMT-S2; MRT51
UniProt ID: (Human) P50135, (Mouse) Q91VF2, (Rat) Q01984
Entrez Gene ID: (Human) 3176, (Mouse) 140483, (Rat) 81676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support