Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METEQPEETF PNTETNGEFG KRPAEDMEEE QAFKRSRNTD EMVELRILLQ SKNAGAVIGK GGKNIKALRT DYNASVSVPD SSGPERILSI SADIETIGEI LKKIIPTLEE GLQLPSPTAT SQLPLESDAV ECLNYQHYKG SDFDCELRLL IHQSLAGGII GVKGAKIKEL RENTQTTIKL FQECCPHSTD RVVLIGGKPD RVVECIKIIL DLISESPIKG RAQPYDPNFY DETYDYGGFT MMFDDRRGRP VGFPMRGRGG FDRMPPGRGG RPMPPSRRDY DDMSPRRGPP PPPPGRGGRG GSRARNLPLP PPPPPRGGDL MAYDRRGRPG DRYDGMVGFS ADETWDSAID TWSPSEWQMA YEPQGGSGYD YSYAGGRGSY GDLGGPIITT QVTIPKDLAG SIIGKGGQRI KQIRHESGAS IKIDEPLEGS EDRIITITGT QDQIQNAQYL LQNSVKQYSG KFF
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene is located in the nucleoplasm and has three repeats of KH domains that binds to RNAs. It is distinct among other hnRNP proteins in its binding preference; it binds tenaciously to poly(C). This protein is also thought to have a role during cell cycle progression. Several alternatively spliced transcript variants have been described for this gene, however, not all of them are fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dC-stretch binding protein; FLJ41122; Heterogeneous nuclear ribonucleoprotein K; hnRNP K; RP11-575L7.1; Transformation up-regulated nuclear protein; transformation upregulated nuclear protein; TUNP
Gene Aliases: AUKS; CSBP; HNRNPK; HNRPK; TUNP
UniProt ID: (Human) P61978
Entrez Gene ID: (Human) 3190
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support