Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: GSKINASKNQ QDDGKMFIGG LSWDTSKKDL TEYLSRFGEV VDCTIKTDPV TGRSRGFGFV LFKDAASVDK VLELKEHKLD GKLIDPKRAK ALKGKEPPKK VFVGGLSPDT SEEQIKEYFG AFGEIENIEL PMDTKTNERR GFCFITYTDE EPVKKLLESR YHQIGSGKCE IKVAQPKEVY RQQQQQQKGG RGAAAGGRGG TRGRGRGQGQ NWNQGFNNYY DQGYGNYNSA YGGDQNYSGY GGYDYTGYNY GNYGYGQGYA DYSGQQSTYG KASRGGGNHQ NNYQPY
hnRNP DL belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two RRM domains that bind to RNAs. Three alternatively spliced transcript variants have been described for this gene. One of the variants is probably not translated because the transcript is a candidate for nonsense-mediated mRNA decay. The protein isoforms encoded by this gene are similar to its family member HNRPD.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A+U-rich element RNA binding factor; AU-rich element RNA-binding factor; Heterogeneous nuclear ribonucleoprotein D-like; hnRNP D-like; hnRNP DL; hnRPD-like protein; JKT41-binding protein; Protein laAUF1
Gene Aliases: AA407431; AA959857; D5Ertd650e; D5Wsu145e; HNRNP; hnRNP DL; hnRNP-DL; HNRNPDL; Hnrnpdl1; HNRPDL; JKTBP; JKTBP2; laAUF1; LGMD1G
UniProt ID: (Human) O14979
Entrez Gene ID: (Human) 9987, (Mouse) 50926
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support