Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: RQHFTIALK EKHITDMVCP ACGRPDLTDD TQLLSYFSTL DIQLRESLEP DAYALFHKKL TEGVLMRDPK FLWCAQCSFG FIYEREQLEA TCPQCHQTFC VRCKRQWEEQ HRGRSCEDFQ NWKRMNDPEY QAQGLAMYLQ ENGIDCPKCK FSYALARGGC MHFHCTQCRH QFCSGCYNAF YAKNKCPEPN CRVKKSLHGH HPRDCLFYLR DWTALRLQKL LQDNNVMFNT EPPAGARAVP GGGCRVIEQK EVPNGLRDEA CGKETPAGYA GLCQAHYKEY LVSLINAHSL DPATLYEVEE LETATERYLH VRPQPLAGED PPAYQARLLQ KLTEEVPLGQ SIPRRRK (258-603 aa encoded by BC017376 )
Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RNF31; FLJ23501; HOIL-1-interacting protein; HOIP; MGC19975; RING finger protein 31; RING-type E3 ubiquitin transferase RNF31; Zinc in-between-RING-finger ubiquitin-associated domain protein
Gene Aliases: HOIP; RNF31; ZIBRA
UniProt ID: (Human) Q96EP0
Entrez Gene ID: (Human) 55072
Molecular Function:
ubiquitin-protein ligase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support