Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
21653-1-AP is a rabbit polyclonal antibody raised against residues near the N-terminus of human HOXB1.
Immunogen sequence: MDYNRMNSF LEYPLCNRGP SAYSAHSAHS APTSFPPSSA QAVDSYASEG RYGGGLSSPA FQQNSGYPAQ QPPSTLGVPF PSSAPSGYAP AACSPSYGPS QYYPLGHSEG DGGYFHPSSY GAQLGGLSDG YGAGGAGPGP YPPQHPPYGN EQTASFAPAY ADLLSEDKE (1-168 aa encoded by BC096192 )
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Acts on the anterior body structures.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box B1; Homeobox protein Hox-2.9; Homeobox protein Hox-2I; Homeobox protein Hox-B1; Hox-29
Gene Aliases: HCFP3; Hox-2.9; HOX2; HOX2I; Hoxb-1; HOXB1
UniProt ID: (Human) P14653, (Mouse) P17919
Entrez Gene ID: (Human) 3211, (Mouse) 15407
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support