Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSISGTLSSY YVDSIISHES EDAPPAKFPS GQYASSRQPG HAEHLEFPSC SFQPKAPVFG ASWAPLSPHA SGSLPSVYHP YIQPQGVPPA ESRYLRTWLE PAPRGEAAPG QGQAAVKAEP LLGAPGELLK QGTPEYSLET SAGREAVLSN QRPGYGDNKI CEGSEDKERP DQTNPSANWL HARSSRKKRC PYTKYQTLEL EKEFLFNMYL TRDRRHEVAR LLNLSERQVK IWFQNRRMKM KKMNKEQGKE
HOXB9 belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. HOXB9 is one of several homeobox HOXB genes located in a cluster on chromosome 17. The exact role of this gene has yet to be determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:5120; homeo box 2E; homeo box B9; Homeobox protein Hox-2.5; Homeobox protein Hox-2E; Homeobox protein Hox-B9; MGC124051; MGC124052; MGC124053; MGC124054
Gene Aliases: HOX-2.5; HOX2; HOX2E; HOXB9
UniProt ID: (Human) P17482
Entrez Gene ID: (Human) 3219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support