Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: DRAPQTLPSP EADALAGSKH KEEKADLDPS NPVANWIHAR STRKKRCPYT
Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 79%; Rabbit: 93%; Rat: 100%; Sheep: 100%
HOXC9 belongs to the homeobox family. The homeobox family is a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms.This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein Hox-3B; Homeobox protein Hox-C9
Gene Aliases: HOX3; HOX3B; HOXC9
UniProt ID: (Human) P31274
Entrez Gene ID: (Human) 3225
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support