Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: FEPHPMASNG GGFASANLQG SPVYVGGNFV ESMAPASGPV FNLGHLSHPS SASVDYSCAA QIPGNHHHGP CDPHPTYTDL SAHHSSQGRL PEAPKLTH
HOXD3 (homeobox D3) belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located at 2q31-2q37 chromosome regions. Deletions that remove the entire HOXD gene cluster or 5' end of this cluster have been associated with severe limb and genital abnormalities. HOXD3 may play a role in the regulation of cell adhesion processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box D3; Homeobox protein Hox-4A; Homeobox protein Hox-D3; Hox-4.1, mouse, homolog of; MGC10470
Gene Aliases: Hox-4.1; HOX1D; HOX4; HOX4A; HOXD3
UniProt ID: (Human) P31249
Entrez Gene ID: (Human) 3232
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support