Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: SH-SY5Y, HepG2, HT-1080, NCI-H460, HeLa, Mouse brain, Mouse liver, Mouse kidney, Rat brain
Immunogen sequence: MGKQNSKLRP EVLQDLRENT EFTDHELQEW YKGFLKDCPT GHLTVDEFKK IYANFFPYGD ASKFAEHVFR TFDTNGDGTI DFREFIIALS VTSRGKLEQK LKWAFSMYDL DGNGYISRSE MLEIVQAIYK MVSSVMKMPE DESTPEKRTD KIFRQMDTNN DGKLSLEEFI RGAKSDPSIV RLLQCDPSSA SQF
Hippocalcin-like protein 1, may be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-binding protein BDR-1; hippocalcin-like 1; hippocalcin-like 1; visinin like 3; neural visinin-like 3; neural visinin-like protein 3; Hippocalcin-like protein 1; HLP2; neural visinin-like 3; neural visinin-like Ca2+-binding protein type 3; Neural visinin-like protein 3; NVL-3; VILIP-3; visinin like 3; Visinin-like protein 3
Gene Aliases: BDR1; HLP2; HPCAL1; NVP-3; Nvp3; VILIP-3; VILIP3; Vnsl3; Vsnl3
UniProt ID: (Human) P37235, (Rat) P62749, (Mouse) P62748
Entrez Gene ID: (Human) 3241, (Rat) 50871, (Mouse) 53602
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support