Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPNYKLTYF NMRGRAEIIR YIFAYLDIQY EDHRIEQADW PEIKSTLPFG KIPILEVDGL TLHQSLAIAR YLTKNTDLAG NTEMEQCHVD AIVDTLDDFM SCFPWAEKKQ DVKEQMFNEL LTYNAPHLMQ DLDTYLGGRE WLIGNSVTWA DFYWEICSTT LLVFKPDLLD NHPRLVTLRK KVQAIPAVAN WIKRRPQTKL (1-199 aa encoded by BC020734)
PGDS is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes. Prostaglandin-D synthase is a sigma class glutathione-S-transferase family member. The enzyme catalyzes the conversion of PGH2 to PGD2 and plays a role in the production of prostanoids in the immune system and mast cells. The presence of this enzyme can be used to identify the differentiation stage of human megakaryocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.5.1.18; EC 5.3.99.2; Glutathione S-transferase; glutathione S-transferase sigma; Glutathione-dependent PGD synthase; glutathione-dependent PGD synthetase; Glutathione-requiring prostaglandin D synthase; GST class-sigma; H-PGDS; Hematopoietic prostaglandin D synthase; hematopoietic prostaglandin D2 synthase; prostaglandin D2 synthase 2, hematopoietic; Prostaglandin-H2 D-isomerase
Gene Aliases: GSTS; GSTS1; GSTS1-1; H-PGDS; HPGDS; PGD2; PGDS; PTGDS2
UniProt ID: (Human) O60760, (Mouse) Q9JHF7
Entrez Gene ID: (Human) 27306, (Mouse) 54486
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support