Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: VAVLLRSDQE PLYPVQVSSA DARLMVFDKT EGTWRLLCSS RSNARVAGLS CEEMGFLRAL THSELDVRTA GANGTSGFFC VDEGRLPHTQ RLLEVISVCD CPRGRFLAAI CQDCGRRKLP VDRIVGGRDT SLGRWPWQVS LRYDGAHLCG GSLLSGDWVL TAAHCFPERN RVLSRWRVFA GAVAQASPHG LQLGVQAVVY HGGYLPFRDP NSEENSNDIA LVHLSSPLPL TEYIQPVCLP AAGQALVDGK ICTVTGWGNT QYYGQQAGVL QEARVPIISN DVCNGADFYG NQIKPKMFCA GYPEGGIDAC QGDSGGPFAC EDSISRTPRW RLCGIVSWGT GCALAQKPGV YTKVSDFREW IFQAIKTHSE ASGMVTQL
Hepsin is a cell surface serine protease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine protease hepsin; testicular tissue protein Li 85; Transmembrane protease serine 1
Gene Aliases: HPN; TMPRSS1
UniProt ID: (Human) P05981
Entrez Gene ID: (Human) 3249
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support