Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGPRT; HGPRTase; HPRT 1; HPRT B; hypoxanthine guanine phosphoribosyl transferase 1; hypoxanthine phosphoribosyl transferase; Hypoxanthine-guanine phosphoribosyltransferase; hypoxanthine-guanine phosphoribosyltransferase 1; testicular tissue protein Li 89
Gene Aliases: C81579; HGPRT; Hgprtase; HPGRT; HPRT; HPRT1
UniProt ID: (Human) B2R8M9, (Mouse) P00493, (Rat) P27605
Entrez Gene ID: (Human) 3251, (Mouse) 15452, (Rat) 24465
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support