Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
May regulate the synthesis and function of lysosomes and of highly specialized organelles, such as melanosomes and platelet dense granules. Regulates intracellular vesicular trafficking in fibroblasts. May be involved in the regulation of general functions of integrins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha integrin binding protein 63; Alpha-integrin-binding protein 63; BLOC-2 complex member HPS5; Hermansky-Pudlak syndrome 5 protein; Hermansky-Pudlak syndrome 5 protein homolog; Ru2; ruby eye 2; Ruby-eye protein 2; Ruby-eye protein 2 homolog
Gene Aliases: Aa2-028; Ab1-018; AI646796; AIBP63; AL022647; BLOC2S2; C85120; haze; HPS5; hz; KIAA1017; maroon; mr; ru-2; Ru2
UniProt ID: (Human) Q9UPZ3, (Mouse) P59438
Entrez Gene ID: (Human) 11234, (Mouse) 246694, (Rat) 308598
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support