Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KFKNSTYSR SSVDVLYTFA NCSGLDLIFG LNALLRTADL QWNSSNAQLL LDYCSSKGYN ISWELGNEPN SFLKKADIFI NGSQLGEDFI QLHKLLRKST FKNAKLYGPD VGQPRRKTAK MLKSFLKAGG EVIDSVTWHH YYLNGRTATR EDFLNPDVLD IFISSVQKVF QVVESTRPGK KVWLGETSSA YGGGAPLLSD TFAAGFMWLD KLGLSARMGI EVVMRQVFFG AGNYHLVDEN FDPLPDYWLS LLFKKLVGTK VLMASVQGSK RRKLRVYLHC TNTDNPRYKE GDLTLYAINL HNVTKYLRLP YPFSNKQVDK YLLRPLGPHG LLSKSVQLNG LTLKMVDDQT LPPLMEKPLR PGSSLGLPAF SYSFFVIRNA KVAACI (159-543 aa encoded by BC051321)
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al. ). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endo-glucoronidase; endoglycosidase heparanase; Heparanase; Heparanase-1; heparanase-like; Hpa1
Gene Aliases: HEP; HPA; HPA1; HPR1; HPSE; HPSE1; HSE1
UniProt ID: (Human) Q9Y251, (Mouse) Q6YGZ1, (Rat) Q71RP1
Entrez Gene ID: (Human) 10855, (Mouse) 15442, (Rat) 64537
Molecular Function:
glycosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support