Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MAFNDCFSLN YPGNPCPGDL IEVFRPGYQH WALYLGDGYV INIAPVDGIP ASFTSAKSVF SSKALVKMQL LKDVVGNDTY RINNKYDETY PPLPVEEIIK RSEFVIGQEV AYNLLVNNCE HFVTLLRYGE GVSEQANRAI STVEFVTAAV GVFSFLGLFP KGQRAKYY
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-REV107 protein-related protein; HRAS-like suppressor 1; HRSL1; phospholipase A; Phospholipase A and acyltransferase 1; Phospholipid-metabolizing enzyme A-C1
Gene Aliases: A-C1; H-REV107; HRASLS; HRASLS1; HRSL1; HSD28; PLAAT1
UniProt ID: (Human) Q9HDD0
Entrez Gene ID: (Human) 57110
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support