Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
H2 Histamine Receptor stimulates adenylate cyclase activity and leads to the release of acids in the stomach. It is also involved in GI motility and intestinal secretion. Treatments for acid-peptic disorders are based on the inhibition of histamine H2 receptor activity. The 3-untranslated region may be involved in differential expression. Histamine H2 receptor has been reported to be expressed in bone marrow, brain, heart, lymph node, nose, placenta, spinal cord, and stomach. An EST has been isolated from a kidney library.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gastric receptor 1; Gastric receptor I; H2R; HH2R; Histamine H2 receptor; histamine receptor
Gene Aliases: H2R; HRH2
UniProt ID: (Human) P25021, (Mouse) P97292
Entrez Gene ID: (Human) 3274, (Rat) 25461, (Mouse) 15466
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support