Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Samples: Mouse testis, Mouse brain, Mouse spleen, Rat testis, Rat brain, Rat spleen, Raji, SH-SY5Y, U-251MG
Immunogen sequence: MERAPPDGPL NASGALAGEA AAAGGARGFS AAWTAVLAAL MALLIVATVL GNALVMLAFV ADSSLRTQNN FFLLNLAISD FLVGAFCIPL YVPYVLTGRW
H3 Histamine Receptor regulates the release of the neurotransmitters norepinephrine and somatostatin. In histamine-containing neurons of the brain, the H3 receptor acts as a presynaptic autoreceptor, while in nonhistamine-containing neurons, in both the central and peripheral nervous systems, it acts as a presynaptic heteroreceptor. Multiple isoforms of H3 receptors are produced by alternative splicing. Histamine H3 receptor has been reported primarily in brain. ESTs have been isolated from brain (normal and cancer), breast cancer, colon cancer, and retinoblastoma libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 97; G-protein coupled receptor 97; H3R; HH3R; Histamine H3 receptor; histamine receptor
Gene Aliases: AW049250; GPCR97; H3R; HH3R; HRH3
UniProt ID: (Human) Q9Y5N1, (Rat) Q9QYN8, (Mouse) P58406
Entrez Gene ID: (Human) 11255, (Rat) 85268, (Mouse) 99296
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support