Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
Recommended positive controls: HCT116.
Predicted reactivity: Mouse (81%), Rat (86%), Bovine (86%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Endoribonuclease responsible for the inhibition of the translation by cleaving mRNA. Inhibits cell-free protein synthesis. Cleaves phosphodiester bonds only in single-stranded RNA.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14.5 kDa translational inhibitor protein; 2-iminobutanoate/2-iminopropanoate deaminase; Heat-responsive protein 12; hp14.5; perchloric acid-soluble protein; Reactive intermediate imine deaminase A homolog; reactive intermediate/imine deaminase A homolog; Translation inhibitor L-PSP ribonuclease; translational inhibitor p14.5; translational inhibitor protein p14.5; UK114 antigen homolog
Gene Aliases: HRSP12; P14.5; PSP; RIDA; UK114
UniProt ID: (Human) P52758
Entrez Gene ID: (Human) 10247
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support