Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MQSPAVLVTS RRLQNAHTGL DLTVPQHQEV RGKMMSGHVE YQILVVTRLA AFKSAKHRPE DVVQFLVSKK YSEIEEFYQK LSSRYAAASL PPLPRKVLFV GESDIRERRA VFNEILRCVS KDAELAGSPE LLEFLGTRSP GAAGLTSRDS SVLDGTDSQT GNDEEAFDFF EEQDQVAEEG PPVQSLKGED AEESLEEEEA LDPLGIMRLV SCC
The protein encoded by this gene shares similarity with mouse Hs1bp3, an Hcls1/Hs1-interacting protein that may be involved in lymphocyte activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HCLS1-binding protein 3; HS1-binding protein 3; HSP1BP-3
Gene Aliases: ETM2; HS1-BP3; HS1BP3
UniProt ID: (Human) Q53T59
Entrez Gene ID: (Human) 64342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support