Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: QIHFVSGERL ITDPAGEMGR VQDFLGIKRF ITDKHFYFNK TKGFPCLKKT ESSLLPRCLG KSKGRTHVQI DPEVIDQLRE FYRPYNIKFY ETVGQDFRWE
Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. This gene is expressed predominantly in brain and may play a role in the nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-OST-2; h3-OST-2; heparan sulfate (glucosamine) 3-O-sulfotransferase 2; heparan sulfate 3-O-sulfotransferase 2; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 2; Heparan sulfate glucosamine 3-O-sulfotransferase 2; heparin-glucosamine 3-O-sulfotransferase
Gene Aliases: 30ST2; 3OST2; HS3ST2; UNQ2442/PRO5004
UniProt ID: (Human) Q9Y278
Entrez Gene ID: (Human) 9956
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support