Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Recommended positive controls: 293T, Raji.
Predicted reactivity: Mouse (82%), Rat (81%), Rhesus Monkey (99%), Bovine (88%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ366L4.2; DnaJ (Hsp40) homolog, subfamily C, member 20; DnaJ homolog subfamily C member 20; HscB iron-sulfur cluster co-chaperone homolog; HscB mitochondrial iron-sulfur cluster co-chaperone; Iron-sulfur cluster co-chaperone protein HscB; J-type co-chaperone HSC20
Gene Aliases: DNAJC20; HSC20; HSCB; JAC1
UniProt ID: (Human) Q8IWL3
Entrez Gene ID: (Human) 150274
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support