Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
May act as a co-chaperone in iron-sulfur cluster assembly in mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ366L4.2; DnaJ (Hsp40) homolog, subfamily C, member 20; DnaJ homolog subfamily C member 20; HscB iron-sulfur cluster co-chaperone homolog; HscB mitochondrial iron-sulfur cluster co-chaperone; Iron-sulfur cluster co-chaperone protein HscB; iron-sulfur cluster co-chaperone protein HscB, mitochondrial; J-type co-chaperone HSC20
Gene Aliases: AI325508; AW049829; DNAJC20; HSC20; HSCB; JAC1; RGD1311005
UniProt ID: (Human) Q8IWL3, (Mouse) Q8K3A0
Entrez Gene ID: (Human) 150274, (Dog) 477531, (Rat) 360826, (Mouse) 100900
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support