Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: EEFRPEMLQG KKVIVTGASK GIGREMAYHL AKMGAHVVVT ARSKETLQKV VSHCLELGAA SAHYIAGTME DMTFAEQFVA QAGKLMGGLD MLILNHITNT SLNLFHDDIH HVRKSMEVNF LSYVVLTVAA LPMLKQSNGS IVVVSSLAGK VAYPMVAAYS ASKFALDGFF SSIRKEYSVS RVNVSITLCV LGLIDTETAM KAVSGIVHMQ AAPKEECALE IIKGGALRQE EVYYDSSLWT TLLIRNPCRK ILEFLYSTSY NMDRFINK; Positive Samples: Mouse liver; Cellular Location: Endoplasmic reticulum membrane, Single-pass type II membrane protein
HSD11B1 is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, HSD11B1 can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 11-beta-HSD1; 11-beta-HSD1A/MCR; 11-beta-hydroxysteroid dehydrogenase 1; 11-beta-hydroxysteroid dehydrogenase/microsomal carbonyl reductase; 11-DH; 11beta-HSD-1; 11beta-HSD1A; 11beta-hydroxysteroid dehydrogenase type 1; 7-oxosteroid reductase; Corticosteroid 11-beta-dehydrogenase isozyme 1; Corticosteroid 11-beta-dehydrogenase, isozyme 1 (11-DH) (11-beta-hydroxysteroid dehydrogenase 1) (11-beta-HSD1) (11beta-HSD1A); liver-type 11-beta-HSD; MGC13539; OTTHUMP00000034650
Gene Aliases: Hsd11; Hsd11b1
UniProt ID: (Mouse) P50172
Entrez Gene ID: (Mouse) 15483
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support