Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: VHTAFMEKVL GSPEEVLDRT DIHTFHRFYQ YLAHSKQVFR EAAQNPEEVA EVFLTALRAP KPTLRYFTTE RFLPLLRMRL DDPSGSNYVT AMHREVF
HSD17B1 is favors the reduction of estrogens and androgens. It also has 20-alpha-HSD activity. It uses preferentially NADH.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E2DH; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; Placental 17-beta-hydroxysteroid dehydrogenase; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1
Gene Aliases: E17KSR; EDH17B1; EDH17B2; EDHB17; HSD17; HSD17B1; SDR28C1
UniProt ID: (Human) P14061
Entrez Gene ID: (Human) 3292
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support