Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IVCSLESFV KLFIPKRRKS VTGEIVLITG AGHGIGRLTA YEFAKLKSKL VLWDINKHGL EETAAKCKGL GAKVHTFVVD CSNREDIYSS AKKVKAEIGD VSILVNNAGV VYTSDLFATQ DPQIEKTFEV NVLAHFWTTK AFLPAMTKNN HGHIVTVASA AGHVSVPFLL AYCSSKFAAV GFHKTLTDEL AALQITGVKT TCLCPNFVNT GFIKNPSTSL GPTLEPEEVV NRLMHGILTE QKMIFIPSSI AFLTTLERIL PERFLAVLKR KISVKFDAVI GYKMKAQ (15-300 aa encoded by BC016367)
Short-chain alcohol dehydrogenases, such as HSD17B11, metabolize secondary alcohols and ketones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17-beta-HSD 11; 17-beta-HSD XI; 17-beta-hydroxysteroid dehydrogenase 11; 17-beta-hydroxysteroid dehydrogenase type XI; 17-beta-hydroxysteroid dehydrogenase XI; 17beta-HSD11; 17betaHSD11; 17betaHSDXI; 17bHSD11; CTCL tumor antigen HD-CL-03; CTCL-associated antigen HD-CL-03; Cutaneous T-cell lymphoma-associated antigen HD-CL-03; dehydrogenase/reductase (SDR family) member 8; Dehydrogenase/reductase SDR family member 8; Estradiol 17-beta-dehydrogenase 11; hydroxysteroid (17-beta) dehydrogenase 11; Retinal short-chain dehydrogenase/reductase 2; retinal short-chain dehydrogenase/reductase SDR2; retSDR2; Short chain dehydrogenase/reductase family 16C member 2; short chain dehydrogenase/reductase family 16C, member 2
Gene Aliases: 17-BETA-HSD11; 17-BETA-HSDXI; 17BHSD11; DHRS8; HSD17B11; PAN1B; PSEC0029; RETSDR2; SDR16C2; SDR2; UNQ207/PRO233
UniProt ID: (Human) Q8NBQ5, (Rat) Q6AYS8, (Mouse) Q3U2P6
Entrez Gene ID: (Human) 51170, (Rat) 289456, (Mouse) 114664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support