Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNIILEILLL LITIIYSYLE SLVKFFIPQR RKSVAGEIVL ITGAGHGIGR QTTYEFAKRQ SILVLWDINK RGVEETAAEC RKLGVTAHAY VVDCSNREEI YRSLNQVKKE VGDVTIVVNN AGTVYPADLL STKDEEITKT FEVNILGHFW ITKALLPSMM ERNHGHIVTV ASVCGHEGIP YLIPYCSSKF AAVGFHRGLT SELQALGKTG IKTSCLCPVF VNTGFTKNPS TRLWPVLETD EVVRSLIDGI LTNKKMIFVP SYINIFLRLQ KFLPERASAI LNRMQNIQFE AVVGHKIKMK
HSD17B13 (Hydroxysteroid 17 Beta Dehydrogenase 13) is a Protein Coding gene. Diseases associated with HSD17B13 include5 Alpha Reductase DeficiencyandMucopolysaccharidosis Iii. Among its related pathways areMetabolismandLipoprotein metabolism. Gene Ontology (GO) annotations related to this gene includeoxidoreductase activity. An important paralog of this gene isHSD17B11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17-beta hydroxysteroid dehydrogenase; 17-beta-HSD 13; 17-beta-hydroxysteroid dehydrogenase 13; Hepatic retinol/retinal dehydrogenase; hydroxysteroid (17-beta) dehydrogenase 13; Short chain dehydrogenase/reductase family 16C member 3; short chain dehydrogenase/reductase family 16C, member 3; Short-chain dehydrogenase/reductase 9
Gene Aliases: HMFN0376; HSD17B13; NIIL497; SCDR9; SDR16C3; UNQ497/PRO1014
UniProt ID: (Human) Q7Z5P4
Entrez Gene ID: (Human) 345275
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support