Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: ATGTRYAGKV VVVTGGGRGI GAGIVRAFVN SGARVVICDK DESGGRALEQ ELPGAVFILC DVTQEDDVKT LVSETIR
Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 71%.
17-beta-hydroxysteroid dehydrogenases, such as HSD17B14, are primarily involved in metabolism of steroids at the C17 position and also of other substrates, such as fatty acids, prostaglandins, and xenobiotics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17-beta-HSD 14; 17-beta-hydroxysteroid dehydrogenase 14; 17-beta-hydroxysteroid dehydrogenase DHRS10; dehydrogenase/reductase (SDR family) member 10; Dehydrogenase/reductase SDR family member 10; hydroxysteroid (17-beta) dehydrogenase 14; retinal short-chain dehydrogenase/reductase 3; Retinal short-chain dehydrogenase/reductase retSDR3; Short chain dehydrogenase/reductase family 47C member 1; short chain dehydrogenase/reductase family 47C, member 1; testicular tissue protein Li 52
Gene Aliases: DHRS10; HSD17B14; retSDR3; SDR3; SDR47C1; UNQ502/PRO474
UniProt ID: (Human) Q9BPX1
Entrez Gene ID: (Human) 51171
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support