Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSTFFSDTAW ICLAVPTVLC GTVFCKYKKS SGQLWSWMVC LAGLCAVCLL ILSPFWGLIL FSVSCFLMYT YLSGQELLPV DQKAVLVTGG DCGLGHALCK YLDELGFTVF AGVLNENGPG AEELRRTCSP RLSVLQMDIT KPVQIKDAYS KVAAMLQDRG LWAVINNAGV LGFPTDGELL LMTDYKQCMA VNFFGTVEVT KTFLPLLRKS KGRLVNVSSM GGGAPMERLA SYGSSKAAVT MFSSVMRLEL SKWGIKVASI QPGGFLTNIA GTSDKWEKLE KDILDHLPAE VQEDYGQDYI LAQRNFLLLI NSLASKDFSP VLRDIQHAIL AKSPFAYYTP GKGAYLWICL AHYLPIGIYD YFAKRHFGQD KPMPRALRMP NYKKKAT
HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1.1.1.239; 17 beta HSD 2; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha HSD; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E2DH; Estradiol 17-beta-dehydrogenase 2; hydroxysteroid (17-beta) dehydrogenase 2; Microsomal 17-beta-hydroxysteroid dehydrogenase; Short chain dehydrogenase/reductase family 9C member 2; short chain dehydrogenase/reductase family 9C, member 2; tes; Testosterone 17-beta-dehydrogenase
Gene Aliases: EDH17B2; HSD17; HSD17B2; SDR9C2
UniProt ID: (Human) P37059
Entrez Gene ID: (Human) 3294
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support