Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLPNTGRLA GCTVFITGAS RGIGKAIALK AAKDGANIVI AAKTAQPHPK LLGTIYTAAE EIEAVGGKAL PCIVDVRDEQ QISAAVEKAI KKFGAYTIAK YGMSMYVLGM AEEFKGEIAV NALWPKTAIH TAAMDMLGGP GIESQCRKVD IIADAAYSIF QKPKSFTGNF VIDENILKEE GIENFDVYAI KPGHPLQPDF FLDEYPEAVS KKVESTGAVP EFKEEKLQLQ PKPRSGAVEE TFRIVKDSLS DDVVKATQAI YLFELSGEDG GTWFLDLKSK GGNVGYGEPS DQADVVMSMT TDDFVKMFSG KLKPTMAFMS GKLKIKGNMA LAIKLEKLMN QMNARL (1-345 aa encoded by B C004331)
HSDL2 is a protein coding gene. Has apparently no steroid dehydrogenase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hydroxysteroid dehydrogenase-like protein 2; Short chain dehydrogenase/reductase family 13C member 1; short chain dehydrogenase/reductase family 13C, member 1
Gene Aliases: 2610207I16Rik; C9orf99; HSDL2; RGD1305387; SDR13C1
UniProt ID: (Human) Q6YN16, (Mouse) Q2TPA8, (Rat) Q4V8F9
Entrez Gene ID: (Human) 84263, (Mouse) 72479, (Rat) 313200
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support