Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKQSSNVPAF LSKLWTLVEE THTNEFITWS QNGQSFLVLD EQRFAKEILP KYFKHNNMAS FVRQLNMYGF RKVVHIDSGI VKQERDGPVE FQHPYFKQGQ DDLLENIKRK VSSSKPEENK IRQEDLTKII SSAQKVQIKQ ETIESRLSEL KSENESLWKE VSELRAKHAQ QQQVIRKIVQ FIVTLVQNNQ LVSLKRKRPL LLNTNGAQKK NLFQHIVKEP TDNHHHKVIF
Prokaryotic and eukaryotic cells respond to thermal and chemical stress by inducing a group of genes collectively designated heat shock genes. In eukaryotes, this gene expression is regulated primarily at the transcription level. Heat shock transcription factors (HSF, also designated HSTF) 1 and 2 are involved in this regulation. HSF1 and HSF2 are upregulated by estrogen at both the mRNA and protein level. HSF1 is normally found as a monomer whose transcriptional activity is repressed by constitutive phosphorylation. Upon activation, HSF1 forms trimers, gains DNA binding activity and is translocated to the nucleus. HSF2 activity is associated with differentiation and development, and, like HSF1, binds DNA as a trimer. Both HSF1 and HSF2 are known to be induced by proteasome inhibitors of the ubiquitin pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock factor protein 2; Heat shock transcription factor 2; HSF 2; HSTF 2; MGC117376; MGC156196; MGC75048
Gene Aliases: HSF 2; HSF2; HSTF 2; HSTF2
UniProt ID: (Human) Q03933
Entrez Gene ID: (Human) 3298
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support