Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: VLMLMSLYNV SINLKGLKYI SESPGFIPLL WWLLSDPDAE VCLHVLRLVQ SVVLEPEVFS KSASEFRSSL PLQRILAMSK SRNPRLQTAA QELLEDLRTL EHNV
HSF2 binding protein (HSF2BP) associates with HSF2. The interaction occurs between the trimerization domain of HSF2 and the amino terminal hydrophilic region of HSF2BP that comprises two leucine zipper motifs. HSF2BP may therefore be involved in modulating HSF2 activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock factor 2-binding protein
Gene Aliases: HSF2BP; MEILB2
UniProt ID: (Human) O75031
Entrez Gene ID: (Human) 11077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support