Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17090 targets HSF4 in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples.
The MA5-17090 immunogen is purified recombinant fragment of human HSF4 expressed in E. Coli.
MA5-17090 detects HSF4 which has a predicted molecular weight of approximately 53kDa.
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AT-HSFB1; ATHSF4; cataract; Heat shock factor protein 4; Heat shock transcription factor 4; hHSF4; HSF 4; HSFB1; HSTF 4; Marner
Gene Aliases: CTM; CTRCT5; HSF4
UniProt ID: (Human) Q9ULV5
Entrez Gene ID: (Human) 3299
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support