Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEALLSTPI NPNNFPAKLW RLVNSPRYRS IRWDGRGEGL LIDQPLFEAE LLSPPGPGGG GGTAGAGAEP ELFKTTSFTS FIRQLNLYGF RKVVLGGPGG GKPAGNGPLH HFHNPHFRRD QPQLLVHLKR LTSANKAKLA AGLEVPCRPP NRFQRLLITS ASAATAPLQH QQPPPPAGPR PEPHGPVAVG QFHRSFRRDS LSPYSCVSTP SHDHSTYPLK GLDRTPVPHR IWQNSLGMHP GQVETSPTFS DKGVPFPVLQ RFPTEVTYTL QPSTTSVHVQ QGPQTMVSSS QKYSNYTPSA QYSQAYYPTA VLQCCSPTHM DALSSCVTPT ASSYAHCNYF QNPSMQSSYP V (1-350 aa encoded by BC033017)
HSF5 may act as a transcriptional factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock factor protein 5; Heat shock transcription factor 5; heat shock transcription factor family member 5; HSF 5; HSTF 5; MLZ-220; Protein expressed in male leptotene and zygotene spermatocytes 220
Gene Aliases: Gm739; HSF 5; HSF5; HSTF 5; HSTF5; MLZ-220
UniProt ID: (Human) Q4G112, (Mouse) Q5ND04
Entrez Gene ID: (Human) 124535, (Mouse) 327992, (Rat) 688111
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support