Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
HSFX1 (Heat Shock Transcription Factor Family, X-Linked 1) is a Protein Coding gene. Diseases associated with HSFX1 include Microsporidiosis. Gene Ontology (GO) annotations related to this gene include DNA-binding transcription factor activity and sequence-specific DNA binding. An important paralog of this gene is HSFX2. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heat shock transcription factor family, X linked 1; Heat shock transcription factor, X-linked; ORC4L; testicular secretory protein Li 21
Gene Aliases: HSFX1; HSFX2; LW-1
UniProt ID: (Human) Q9UBD0
Entrez Gene ID: (Human) 100506164
Molecular Function: winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support