Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAHVSSETQD VSPKDELTAS EASTRSPLCE HTFPGDSDLR SMIEEHAFQV LSQGSLLESP SYTVCVSEPD KDDDFLSLNF PRKLWKIVES DQFKSISWDE
This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more telomeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq, Jul 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock transcription factor 2-like protein; Heat shock transcription factor, Y-linked; HSF2-like
Gene Aliases: HSF2L; HSFY; HSFY1; HSFY2
UniProt ID: (Human) Q96LI6
Entrez Gene ID: (Human) 159119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support