Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.1.1.79; Hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; lipase, hormone-sensitive; LIPS; Monoacylglycerol lipase LIPE; REH; Retinyl ester hydrolase
Gene Aliases: AOMS4; FPLD6; HSL; LHS; LIPE
UniProt ID: (Human) Q05469
Entrez Gene ID: (Human) 3991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support