Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody detects the 65 kDa protein (Erythrocyte 65 kDa protein, P65).
Immunogen sequence: PGGSPTNWT PEAVVMLGTT ASRVTGESCE IQVHPMFEPS QVYSDYRPGL VLPEEAHYFI PQEAVYVAGV HYQARVAEQY EGIPYNSSVL SSPMKQIPEQ KPVQGGPTSS SVFEFPSGQA FLVGHLQNLR LDSGLGPGSP LSSISAPIST DATRLKFHPV FVPHSAPAVL THNNESRSNC VFEFHVHTPS SSSGEGGGIL PQRVYRNRQV AVDLNQEELP PQSVGLHGYL QPVTEEKHNY HAPELTVSVV EPIGQNWPIG SPEYSSDSSQ ITSSDPSDFQ SPPPTGGAAA PFGSDVSMPF IHLPQTVLQE SPLFFCFPQG TTSQQVLTAS FSSGGSALHP QVIGKLPQLF (869-1120 aa encoded by BC130469)
WNK1 serine/threonine kinase is a member of the WNK subfamily. WNK1 may be a key regulator of blood pressure by controlling the transport of sodium and chloride ions. Mutations in this gene have been associated with pseudohypoaldosteronism type II and hereditary sensory neuropathy type II. WNK plays an important role in the regulation of electrolyte homeostasis, cell signaling, survival, and proliferation. Also, acts as an activator and inhibitor of sodium-coupled chloride cotransporters and potassium-coupled chloride cotransporters respectively. Activates SCNN1A, SCNN1B, SCNN1D, and SGK1. WNK localizes to a segment of kidney involved in salt, potassium, and pH homeostasis, the distal nephron. Mutations in the WNK gene result in Pseudohypaldosteronism and Neuropathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Erythrocyte 65 kDa protein; hWNK1; Kinase deficient protein; MGC163339; MGC163341; p65; prostate-derived sterile 20-like kinase; Protein kinase lysine-deficient 1; Protein kinase with no lysine 1; protein phosphatase 1, regulatory subunit 167; Serine/threonine-protein kinase WNK1; serine/threonine-protein kinase WNK1 1; serine/threonine-protein kinase WNK1 2; With no K; WNK lysine deficient protein kinase 1 isoform
Gene Aliases: 6430573H23Rik; EG406236; HSAN2; HSN2; KDP; KIAA0344; mKIAA0344; p65; PPP1R167; PRKWNK1; PSK; WNK1
UniProt ID: (Human) O15052, (Mouse) P83741
Entrez Gene ID: (Human) 65125, (Mouse) 232341
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support